missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human HMGB2 (aa 164-187) Control Fragment Recombinant Protein
Recombinant Protein
Supplier: Invitrogen™ RP103150
Description
Highest antigen sequence indentity to the following orthologs: Mouse (100%), Rat (100%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-62773 (PA5-62773. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
HMGB2 encodes a member of the non-histone chromosomal high mobility group protein family. The proteins of this family are chromatin-associated and ubiquitously distributed in the nucleus of higher eukaryotic cells. In vitro studies have demonstrated that this protein is able to efficiently bend DNA and form DNA circles. These studies suggest a role in facilitating cooperative interactions between cis-acting proteins by promoting DNA flexibility. This protein was also reported to be involved in the final ligation step in DNA end-joining processes of DNA double-strand breaks repair and V(D)J recombination.Specifications
| P26583 | |
| Blocking Assay, Control | |
| 3148 | |
| 100 μL | |
| C80539; high mobility group box 2; high mobility group protein 2; High mobility group protein B2; high-mobility group (nonhistone chromosomal) protein 2; high-mobility group box 2; HMG 2; HMG B2; Hmg2; HMG-2; HMGB2; HMG-B2; HMGB-2 | |
| Hmgb2 | |
| Human | |
| His-ABP-tag | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ≥5.0 mg/mL | |
| 1 M urea, PBS with no preservative; pH 7.4 | |
| Human HMGB2 (aa 164-187) Control Fragment | |
| RUO | |
| HMGB2 | |
| Unconjugated | |
| Recombinant | |
| AKGKSEAGKKGPGRPTGSKKKNEP | |
| E. coli | |
| >80% by SDS-PAGE and Coomassie blue staining |