missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human HLCS (aa 130-257) Control Fragment Recombinant Protein

Catalog No. RP91775
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP91775 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP91775 Supplier Invitrogen™ Supplier No. RP91775
Only null left

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (74%), Rat (74%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-53599 (PA5-53599. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

The protein encoded by this gene shares strong similarity with Saccharomyces cerevisiae Cdc23, a protein essential for cell cycle progression through the G2/M transition. This protein is a component of anaphase-promoting complex (APC), which is composed of eight protein subunits and highly conserved in eukaryotic cells. APC catalyzes the formation of cyclin B-ubiquitin conjugate that is responsible for the ubiquitin-mediated proteolysis of B-type cyclins. This protein and 3 other members of the APC complex contain the TPR (tetratricopeptide repeat), a protein domain important for protein-protein interaction.

Specifications

Accession Number P50747
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 3141
Name Human HLCS (aa 130-257) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 410I21.SP6; biotin- [propriony-Coenzyme A-carboxylase (ATP-hydrolysing)] ligase; Biotin apo-protein ligase; biotin--[acetyl-CoA-carboxylase] ligase; Biotin--[methylcrotonoyl-CoA-carboxylase] ligase; biotin--[methylmalonyl-CoA-carboxytransferase] ligase; Biotin--[propionyl-CoA-carboxylase [ATP-hydrolyzing]] ligase; Biotin--protein ligase; D16Jhu34; HCS; Hlcs; Holocarboxylase synthetase; holocarboxylase synthetase (biotin- [propriony-Coenzyme A-carboxylase (ATP-hydrolysing)] ligase); holocarboxylase synthetase (biotin-(proprionyl-CoA-carboxylase (ATP-hydrolysing)) ligase); holocarboxylase synthetase (biotin-(proprionyl-Coenzyme A-carboxylase (ATP-hydrolysing)) ligase); RGD1565360
Common Name HLCS
Gene Symbol HLCS
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence DLPYDYSSSLESVADETSPEREGRRVNLTGKAPNILLYVGSDSQEALGRFHEVRSVLADCVDIDSYILYHLLEDSALRDPWTDNCLLLVIATRESIPEDLYQKFMAYLSQGGKVLGLSSSFTFGGFQV
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less