missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human HEYL (aa 1-51) Control Fragment Recombinant Protein

Catalog No. RP108673
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP108673 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP108673 Supplier Invitrogen™ Supplier No. RP108673

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (86%), Rat (86%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-85014. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

This gene encodes a member of the hairy and enhancer of split-related (HESR) family of basic helix-loop-helix (bHLH)-type transcription factors. The sequence of the encoded protein contains a conserved bHLH and orange domain, but its YRPW motif has diverged from other HESR family members. It is thought to be an effector of Notch signaling and a regulator of cell fate decisions. Alternatively spliced transcript variants have been found, but their biological validity has not been determined.

Specifications

Accession Number Q9NQ87
Concentration ≥5.0 mg/mL
For Use With (Application) Neutralization, Control
Formulation PBS, 1M urea with no preservative; pH 7.4
Gene ID (Entrez) 26508
Name Human HEYL (aa 1-51) Control Fragment
pH Range 7.4
Purification Method Purified
Quantity 100 μL
Storage Requirements -20°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias bHLHb33; Class B basic helix-loop-helix protein 33; hairy and enhance of split related 3; hairy and enhancer of split-related protein 3; hairy/enhancer-of-split related with YRPW motif 3; hairy/enhancer-of-split related with YRPW motif-like; hairy/enhancer-of-split related with YRPW motif-like protein; Hairy-related transcription factor 3; hes related family bHLH transcription factor with YRPW motif-like; hesr3; hes-related family bHLH transcription factor with YRPW motif-like; HEY3; HEYL; HEY-like protein; hHeyL; hHRT3; Hrt3; HRT-3; mHRT3
Common Name HEYL
Gene Symbol HEYL
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence MKRPKEPSGSDGESDGPIDVGQEGQLSQMARPLSTPSSSQMQARKKHRGII
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less