missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human HB9 (aa 6-43) Control Fragment Recombinant Protein

Catalog No. RP107833
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP107833 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP107833 Supplier Invitrogen™ Supplier No. RP107833

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (84%), Rat (84%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-67195 (PA5-67195. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Homeobox protein HB9 is expressed in lymphoid and pancreatic tissues. It is reported to be Putative transcription factor involved in pancreas development and function. Defects in this gene causes Currarino syndrome. The triad of a presacral tumor, sacral agenesis and anorectal malformation constitutes the Currarino syndrome which is caused by dorsal-ventral patterning defects during embryonic development. It is also reported that HB9 is strongly expressed in epidermal basal cells of the outer scale face in tarsometatarsal scale skin.

Specifications

Accession Number P50219
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 3110
Name Human HB9 (aa 6-43) Control Fragment
Quantity 100 μL
Gene Alias HB9; Hlxb9; homeo box HB9; homeobox gene HB9; homeobox HB9; homeobox protein HB9; homeodomain protein Hb9; HOX SCRA1; HOXHB9; LOW QUALITY PROTEIN: motor neuron and pancreas homeobox protein 1; MNR2; MNX1; motor neuron and pancreas homeobox 1; motor neuron and pancreas homeobox protein 1; SCRA1; zgc:112174
Common Name HB9
Gene Symbol MNX1
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence NFRIDALLAVDPPRAASAQSAPLALVTSLAAAASGTGG
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less