missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human HAUS5 (aa 115-225) Control Fragment Recombinant Protein

Catalog No. RP103652
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP103652 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP103652 Supplier Invitrogen™ Supplier No. RP103652
Only null left

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (71%), Rat (71%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-64319 (PA5-64319. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

The function of this protein has not been specifically defined.

Specifications

Accession Number O94927
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 23354
Name Human HAUS5 (aa 115-225) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 2310022K01Rik; dgt5; HAUS augmin like complex subunit 5; HAUS augmin-like complex subunit 5; HAUS augmin-like complex, subunit 5; HAUS5; KIAA0841
Common Name HAUS5
Gene Symbol HAUS5
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence ARQHTQDTQRRALLLRAQAGAMRRQQHTLRDPMQRLQNQLRRLQDMERKAKVDVTFGSLTSAALGLEPVVLRDVRTACTLRAQFLQNLLLPQAKRGSLPTPHDDHFGTSYQ
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less