missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human HAL (aa 263-360) Control Fragment Recombinant Protein

Catalog No. RP97641
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP97641 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP97641 Supplier Invitrogen™ Supplier No. RP97641
Only null left

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (95%), Rat (95%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-58435 (PA5-58435. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

HAL is a cytosolic enzyme catalyzing the first reaction in histidine catabolism, the nonoxidative deamination of L-histidine to trans-urocanic acid. HAL defects cause histidinemia which is characterized by increased histidine and histamine and decreased urocanic acid in body fluids Histidine ammonia-lyase is a cytosolic enzyme catalyzing the first reaction in histidine catabolism, the nonoxidative deamination of L-histidine to trans-urocanic acid. Histidine ammonia-lyase defects cause histidinemia which is characterized by increased histidine and histamine and decreased urocanic acid in body fluids.

Specifications

Accession Number P42357
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 3034
Name Human HAL (aa 263-360) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias hal; HIS; histidase; histidine ammonia lyase; histidine ammonia-lyase; histidinemia; Hsd; HSTD; Huth; zgc:136980
Common Name HAL
Gene Symbol HAL
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence LALGLVGEGKMWSPKSGWADAKYVLEAHGLKPVILKPKEGLALINGTQMITSLGCEAVERASAIARQADIVAALTLEVLKGTTKAFDTDIHALRPHRG
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less