missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human GRXCR2 (aa 11-80) Control Fragment Recombinant Protein

Catalog No. RP100313
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP100313 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP100313 Supplier Invitrogen™ Supplier No. RP100313

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (91%), Rat (91%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-63718 (PA5-63718. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

This gene encodes a protein containing a glutaredoxin domain, which functions in protein S-glutathionylation. A mutation in this gene was found in a family with autoosomal recessive nonsyndromic sensorineural deafness-101.

Specifications

Accession Number A6NFK2
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 643226
Name Human GRXCR2 (aa 11-80) Control Fragment
Quantity 100 μL
Gene Alias DFNB101; glutaredoxin and cysteine rich domain containing 2; glutaredoxin domain-containing cysteine-rich protein 1-like protein; Glutaredoxin domain-containing cysteine-rich protein 2; glutaredoxin, cysteine rich 2; GRXCR1-like protein; GRXCR2
Common Name GRXCR2
Gene Symbol GRXCR2
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence KSDGKPRKVRFKISSSYSGRVLKQVFEDGQELESPKEEYPHSFLQESLETMDGVYGSGEVPRPQMCSPKL
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less