missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Human Growth Hormone Recombinant Protein, PeproTech®
Recombinant Protein
$125.66 - $1241.41
Specifications
| Accession Number | P01241 |
|---|---|
| For Use With (Application) | Functional Assay |
| Formulation | protein with no preservative |
| Gene ID (Entrez) | 2688 |
| Molecular Weight (g/mol) | 22.3 kDa |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
10040100UG
|
Gibco™
10040100UG |
100 μg |
Each for $335.10
|
|
|||||
1004010UG
|
Gibco™
1004010UG |
10 μg |
Each for $125.66
|
|
|||||
100401MG
|
Gibco™
100401MG |
1 mg |
Each for $1,241.41
|
|
|||||
10040250UG
|
Gibco™
10040250UG |
250 μg |
Each for $435.06
|
|
|||||
10040500UG
|
Gibco™
10040500UG |
500 μg |
Each for $746.37
|
|
|||||
1004050UG
|
Gibco™
1004050UG |
50 μg |
Each for $302.74
|
|
|||||
Description
Recombinant Human Growth Hormone is a 22.3 kDa, single, non-glycosylated polypeptide chain containing 192 amino acid residues. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis
Human growth hormone (hGH) is a member of the somatotropin/prolactin family of hormones which play an important role in growth control. The GH1 gene, along with 4 additional related genes, is located at the growth hormone locus on chromosome 17. The five genes share a remarkably high degree of sequence identity. Alternative splicing results in 5 isoforms ranging from approximately 19 kDa to 24 kDa. HGH is produced by the somatotroph cells in the pituitary. It acts through binding to membrane receptors that belong to the cytokine receptor subfamily. Its major role in stimulating body growth is to stimulate the liver and other tissues to secrete IGF-1. It stimulates both the differentiation and proliferation of myoblasts. It also stimulates amino acid uptake and protein synthesis in muscle and other tissues. Mutations or deletions in the gene lead to growth hormone deficiency and short stature.Specifications
| P01241 | |
| protein with no preservative | |
| 22.3 kDa | |
| E. coli | |
| <1 EU/ μg | |
| Growth Hormone | |
| Determined by its ability to stimulate the proliferation of rat Nb2-11 cells. The expected ED50 is = 0.05 ng/ml, corresponding to a specific activity of = 2 x 10^7 units/mg | |
| Recombinant | |
| -20°C | |
| Lyophilized |
| Functional Assay | |
| 2688 | |
| Human Growth Hormone | |
| RUO | |
| dog growth hormone; GH; Gh1; GHB1; GHB5; GHN; GH-N; Growth hormone; growth hormone 1; growth hormone 12; growth hormone B1; growth hormone B5; growth hormone precursor; HGH; hGH-N; IGHD1B; pituitary growth hormone; RNGHGP; Somatotropin | |
| Gh1 | |
| Unconjugated | |
| MFPTIPLSRL FDNAMLRAHR LHQLAFDTYQ EFEEAYIPKE QKYSFLQNPQ TSLCFSESIP TPSNREETQQ KSNLELLRIS LLLIQSWLEP VQFLRSVFAN SLVYGASDSN VYDLLKDLEE GIQTLMGRLE DGSPRTGQIF KQTYSKFDTN SHNDDALLKN YGLLYCFRKD MDKVETFLRI VQCRSVEGSC GF | |
| E. coli | |
| ≥ 98% by SDS-PAGE gel and HPLC analyses. |