Learn More
Invitrogen™ Human GRO beta (CXCL2) ELISA Development Kit (ABTS), PeproTech®
ELISA
Supplier: Invitrogen™ 900K120K
Description
Produced by activated monocytes and neutrophils and expressed at sites of inflammation. Hematoregulatory chemokine, which, in vitro, suppresses hematopoietic progenitor cell proliferation. GRO-beta(5-73) shows a highly enhanced hematopoietic activity.Specifications
| P19875 | |
| Lot-specific | |
| ELISA | |
| 2920 | |
| Capture Antibody, Detection Antibody, Protein Standard, HRP-conjugate | |
| 10 x 96 Tests | |
| Refer to product documentation for component specific storage temperature. | |
| 1 hrs 45 mins |
| Lot-specific | |
| HRP | |
| Colorimetric Microplate Reader | |
| Cxcl2 | |
| HRP | |
| RUO | |
| CXCL2 | |
| 24 hrs 30 mins |