missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human GRO alpha (CXCL1) ELISA Kit

Catalog No. 5018001
Change view
Click to view available options
Quantity:
96 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
5018001 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 5018001 Supplier Invitrogen™ Supplier No. BMS2122

ELISA

The Human GRO alpha (CXCL1) (Hu GROα) ELISA quantitates Hu GROα in human serum, plasma, buffered solution, or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu GROα.

CXCL1, CXCL2, and CXCL3, also known respectively as GRO alpha, GRO beta (MIP-2 alpha) and GRO gamma (MIP-2 beta), are members of the CXC subfamily of chemokines. Mature CXCL1/2/3 proteins bind with high affinity to the IL-8 receptor type B and are potent neutrophil attractants and activators. In vitro, the processed forms GRO-alpha(4-73), GRO-alpha(5-73) and GRO-alpha(6-73) show a 30-fold higher chemotactic activity.

Specifications

Accession Number P01589
Assay Range 16 to 1,000 pg/mL
Assay Sensitivity 0.27 ng/mL
Conjugate HRP
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
Target Species Human
For Use With (Application) ELISA
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 2919
Gene Symbol CD25,IDDM10,IL2R,IMD41,TCGFR,p55
Interassay CV 9.8%
Intraassay CV 6.7
Kit Contents Pre-coated 96 well plate, Standard, Sample Diluent, Assay Buffer concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Quantity 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Storage Requirements 2°C to 8°C
Synonym CD25 antigen, TAC, Interleukin-2 receptor alpha
Target CXCL1 (GRO alpha)
Test Time 1 hrs 20 mins
Total Assay Time 3 hrs 30 mins
Show More Show Less