missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human Granzyme B ELISA Kit

Catalog No. BMS20272TEN
Change view
Click to view available options
Quantity:
10 x 96 Tests
96 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BMS20272TEN 10 x 96 Tests
BMS20272 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BMS20272TEN Supplier Invitrogen™ Supplier No. BMS20272TEN
Only null left

ELISA

Granzyme B is a member of the granzyme serine protease family, and is found in the granules of cytotoxic T cells and NK cells. Granzyme B has been described as CGL1 (cathepsin G-like-1), a serine protease expressed only in cytotoxic T-lymphocytes after cell activation, and CTLA-1 (cytotoxic T lymphocyte-associated serine esterase 1) based on identification of mRNA in various cytotoxic T cells, but not observed in non-cytotoxic lymphoid cells. Granzyme B is crucial for the rapid induction of target cell death by apoptosis, induced by interaction with cytotoxic T cells. The receptor involved in this process has been identified as mannose 6-phosphate receptor which functions as a death receptor for Granzyme B during cytotoxic T cell-induced apoptosis. Granzyme B enters target cells to cleave caspase-3 and initiate the caspase cascade leading to DNA fragmentation and apoptosis. Granzyme B can also act through a mitochondrial apoptosis pathway by cleaving the Bid protein. Granzymes are neutral serine proteases, which are stored in specialized lytic granules of cytotoxic T lymphocytes (CTLs) and in natural killer (NK) cells. A number of granzymes (A to G) have been isolated and cloned from mouse CTLs and NK cells, however in man, fewer have been cloned and identified.

Specifications

Accession Number P10144
Assay Range 0.7 to 480 pg/mL
Assay Sensitivity <0.43 pg/mL
Conjugate HRP
Product Type ELISA
Sample Type Serum, Supernatant
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 3002
Gene Symbol GZMB
Interassay CV 6.8%
Intraassay CV 6.2
Kit Contents Pre-coated 96 well plate, Standard, Detection antibody (monoclonal antibody to human Granzyme B), Biotin-Conjugate anti-human Granzyme B monoclonal antibody, Streptavidin-HRP, human Granzyme B Standard lyophilized, Wash Buffer Concentrate 20x, Dilution Buffer Concentrate 5x, Substrate Solution, Stop Solution, Adhesive Films
Label or Dye HRP
Quantity 10 x 96 Tests
Regulatory Status RUO
Sample Volume Serum 50 μL, Supernatant 50 μL
Storage Requirements 2°C to 8°C
Target Granzyme B
Test Time 1 hrs
Total Assay Time 2 hrs 30 mins
Show More Show Less