missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human GPRIN3 (aa 110-203) Control Fragment Recombinant Protein

Catalog No. RP100266
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP100266 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP100266 Supplier Invitrogen™ Supplier No. RP100266

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (43%), Rat (43%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibodies, PA5-65350 (PA5-65350, PA5-63465 (PA5-63465. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

May be involved in neurite outgrowth.

Specifications

Accession Number Q6ZVF9
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 285513
Name Human GPRIN3 (aa 110-203) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias C030038J10Rik; C730021L23; FLJ42625; G protein-regulated inducer of neurite outgrowth 3; GPRIN family member 3; Gprin3; GRIN3; KIAA2027; RGD1565690
Common Name GPRIN3
Gene Symbol Gprin3
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence ASAPSSAAGRDLIHTPLTMPANQHTCQSIPGDQPNAITSSMPEDSLMRSQRTSNREQPEKPSCPVGGVLSSSKDQVSCEFPSPETIQGTVQTPV
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less