missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human GPAM (aa 94-175) Control Fragment Recombinant Protein

Catalog No. RP97073
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP97073 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP97073 Supplier Invitrogen™ Supplier No. RP97073

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (89%), Rat (89%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-61218 (PA5-61218. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Glycerol-3-phosphate acyltransferase 1 (GPAT1), one of four known GPAT isoforms, is located on the mitochondrial outer membrane, allowing reciprocal regulation with carnitine palmitoyltransferase-1. It is thought to be critical for the development of hepatic steatosis; steatosis triggered by GPAT1 overexpression leads to hepatic and possibly peripheral insulin resistance. GPAT1 is transcriptionally upregulated by insulin and sterol regulatory element binding protein (SREBP-1) and downregulated by AMP-activated protein kinase. Mice deficient in GPAT1 exhibit decreased triacylglycerol (TAG) in cardiomyocytes even in high-fat diets, suggesting that GPAT1 contributes significantly to TAG accumulation in heart tissue during lipogenic or high fat diets. At least two isoforms of GPAT1 are known to exist.

Specifications

Accession Number Q9HCL2
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 57678
Name Human GPAM (aa 94-175) Control Fragment
Quantity 100 μL
Gene Alias glycerol 3-phosphate acyltransferase, mitochondrial; glycerol-3-phosphate acyltransferase 1, mitochondrial; glycerol-3-phosphate acyltransferase, mitochondrial; GPAM; GPAT; GPAT1; GPAT-1; KIAA1560; P90; RP11-426E5.2
Common Name GPAM
Gene Symbol Gpam
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence INETHTRHRGWLARRLSYVLFIQERDVHKGMFATNVTENVLNSSRVQEAIAEVAAELNPDGSAQQQSKAVNKVKKKAKRILQ
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less