missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human GNG11 (aa 20-43) Control Fragment Recombinant Protein
Recombinant Protein
Supplier: Invitrogen™ RP100041
Description
Highest antigen sequence indentity to the following orthologs: Mouse (100%), Rat (100%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-60963 (PA5-60963. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
Guanine nucleotide-binding proteins (G proteins) are involved as a modulator or transducer in various transmembrane signaling systems. The beta and gamma chains are required for the GTPase activity, for replacement of GDP by GTP, and for G protein-effector interaction.Specifications
| P61952 | |
| Blocking Assay, Control | |
| 2791 | |
| 100 μL | |
| 0610037B21Rik; G protein gamma-11 subunit; G protein subunit gamma 11; GNG11; GNGT11; guanine nucleotide binding protein (G protein), gamma 11; guanine nucleotide binding protein gamma subunit 11; guanine nucleotide-binding protein G(I)/G/G(O) gamma-11 subunit; guanine nucleotide-binding protein G(I)/G/G(O) subunit gamma-11 | |
| GNG11 | |
| Human | |
| His-ABP-tag | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ≥5.0 mg/mL | |
| 1 M urea, PBS with no preservative; pH 7.4 | |
| Human GNG11 (aa 20-43) Control Fragment | |
| RUO | |
| GNG11 | |
| Unconjugated | |
| Recombinant | |
| EQLRKEVKLQRQQVSKCSEEIKNY | |
| E. coli | |
| >80% by SDS-PAGE and Coomassie blue staining |