missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human GLTPD2 (aa 160-186) Control Fragment Recombinant Protein

Catalog No. RP101766
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP101766 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP101766 Supplier Invitrogen™ Supplier No. RP101766

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (63%), Rat (63%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-63551 (PA5-63551. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

GLTPD2 (Glycolipid Transfer Protein Domain Containing 2) is a Protein Coding gene. Gene Ontology (GO) annotations related to this gene include glycolipid binding and glycolipid transporter activity.

Specifications

Accession Number A6NH11
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 388323
Name Human GLTPD2 (aa 160-186) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias C730027E14Rik; Gltpd2; glycolipid transfer protein domain containing 2; glycolipid transfer protein domain-containing protein 2; RGD1560459
Common Name GLTPD2
Gene Symbol GLTPD2
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence RAGLLEQPGAAPRDPTRSSGSRTLLLL
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less