missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human GGA2 (aa 299-383) Control Fragment Recombinant Protein

Catalog No. RP104770
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP104770 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP104770 Supplier Invitrogen™ Supplier No. RP104770
Only null left

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (60%), Rat (60%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-65356 (PA5-65356. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

This gene encodes a member of the Golgi-localized, gamma adaptin ear-containing, ARF-binding (GGA) family. This family includes ubiquitous coat proteins that regulate the trafficking of proteins between the trans-Golgi network and the lysosome. These proteins share an amino-terminal VHS domain which mediates sorting of the mannose 6-phosphate receptors at the trans-Golgi network. They also contain a carboxy-terminal region with homology to the ear domain of gamma-adaptins. This family member may play a significant role in cargo molecules regulation and clathrin-coated vesicle assembly.

Specifications

Accession Number Q9UJY4
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 23062
Name Human GGA2 (aa 299-383) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 1200007E24Rik; ADP-ribosylation factor binding protein 2; ADP-ribosylation factor-binding protein GGA2; gam; Gamma-adaptin-related protein 2; Gga2; golgi associated, gamma adaptin ear containing, ARF binding protein 2; golgi-associated, gamma adaptin ear containing, ARF binding protein 2; golgi-localized, gamma ear-containing, ARF-binding protein 2; Kiaa1080; mKIAA1080; VEAR; VHS domain and ear domain of gamma-adaptin; VHS domain and ear domain-containing protein
Common Name GGA2
Gene Symbol GGA2
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence ANDLLTQGVLLYKQVMEGRVTFGNRVTSSLGDIPVSRVFQNPAGCMKTCPLIDLEVDNGPAQMGTVVPSLLHQDLAALGISDAPV
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less