missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human GDF-15 Quantikine ELISA Kit

Catalog No. DGD150
Change view
Click to view available options
Sufficient For:
1 x 96-well Plate
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sufficient For
DGD150 1 x 96-well Plate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. DGD150 Supplier R&D Systems™ Supplier No. DGD150

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Human GDF-15 Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Human GDF-15 Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies human GDF-15 in cell culture supernates (50 ul), serum (13 ul), edta plasma (13 ul), heparin plasma (13 ul), urine (10 ul).

Specifications

Assay Range 23.40 - 1500pg/mL
Assay Sensitivity 4.39pg/mL
Conjugate HRP
Sample Type Cell Culture Supernatant, Plasma (EDTA, Heparin), Serum, Urine
Specificity Natural and Recombinant Human GDF-15
Sufficient For 1 x 96-well Plate
For Use With (Application) ELISA
Gene ID (Entrez) 9518
Sample Volume Cell Culture Supernates (50uL),Serum (13uL),EDTA Plasma (13uL),Heparin Plasma (13uL),Urine (10uL)
Storage Requirements Store the unopened product at 2° to 8°C. Do not use past expiration date.
Synonym GDF15, growth differentiation factor 15, growth/differentiation factor 15, Macrophage inhibitory cytokine 1, MIC-1, MIC-1 NSAID-activated gene 1 protein, MIC1Prostate differentiation factor, NAG-1, NAG-1 NSAID-regulated gene 1 protein, NSAID (nonsteroidal inflammatory drug)-activated protein 1, PDF, PDFGDF-15, PLAB, PLABNRG-1, Placental bone morphogenetic protein, Placental TGF-beta, PTGF-beta, PTGFBPTGF-beta
Show More Show Less