missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human gamma-Thrombin Native Protein

Catalog No. PIRP43107
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIRP43107 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIRP43107 Supplier Invitrogen™ Supplier No. RP43107

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Native Protein

RP-43107 contains native human g-Thrombin protein. RP-43107 may be used as a positive control in many immunological assays. The product is of human blood/plasma origin. Although the starting material was tested prior to initiation of the manufacturing process, and was found negative or nonreactive for anti-HIV-1/2, HIV-1 antigen(s), HBsAg, STS, anti-HCV, anti-HBcore and anti-HTLV I & II, extreme caution should be used when handling this material as there is a margin of error in all tests. This product is to be used for IN VITRO research purposes only, and is not intended for clinical or diagnostic use!.

Specifications

Accession Number P00734
Concentration 2.0 mg/mL
For Use With (Application) Neutralization
Formulation 0.1M sodium phosphate with 0.1% PEG and no preservative; pH 6.5
Gene ID (Entrez) 2147
Molecular Weight (g/mol) 35.4 kDa
Name Human gamma-Thrombin
pH Range 6.5
Purification Method SDS-PAGE
Quantity 100 μg
Source Human
Storage Requirements -80°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias Activation peptide fragment 1; Activation peptide fragment 2; Coagulation factor II; coagulation factor II (thrombin); coagulation factor II, thrombin; F2; prepro-coagulation factor II; prothrombin; prothrombin B-chain; PT; RPRGL2; serine protease; THPH1; Thrombin heavy chain; Thrombin light chain
Common Name gamma-Thrombin
Gene Symbol F2
Product Type Protein
Conjugate Unconjugated
Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Form Liquid
Protein Subtype Native
Show More Show Less