missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human Galectin-8 ELISA Kit

Catalog No. p-7219391
Encompass
Change view
Click to view available options
Quantity:
5 x 96 Tests
96 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EH205RBX5 5 x 96 Tests
EH205RB 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. EH205RBX5 Supplier Invitrogen™ Supplier No. EH205RBX5
Only null left

ELISA

Human Galectin-8 quantitates human Galectin-8 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human Galectin-8.

This gene encodes a member of the galectin family. Galectins are beta-galactoside-binding animal lectins with conserved carbohydrate recognition domains. The galectins have been implicated in many essential functions including development, differentiation, cell-cell adhesion, cell-matrix interaction, growth regulation, apoptosis, and RNA splicing. This gene is widely expressed in tumoral tissues and seems to be involved in integrin-like cell interactions. Alternatively spliced transcript variants encoding different isoforms have been identified.

Specifications

Accession Number 1200015E08Rik; 30 kDa S-type lectin; AI326142; D13Ertd524e; Gal-8; galectin 8; Galectin-8; galectin-8 g; galectin-8 L; lectin, galactose binding, soluble 5; lectin, galactose binding, soluble 8; lectin, galactoside binding soluble 8; lectin, galactoside-binding, soluble, 8; Lgals8; Lgals-8; PCTA1; PCTA-1; Po66 carbohydrate binding protein; Po66 carbohydrate-binding protein; Po66-CBP; prostate carcinoma tumor antigen 1; RL-30; RP11-385F5.1
Assay Range 0.16 to 40 ng/mL
Assay Sensitivity 0.16 ng/mL
Conjugate HRP
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 3964
Gene Symbol Lgals8
Interassay CV <12%
Intraassay CV <10
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Quantity 5 x 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Storage Requirements 2°C to 8°C
Target Galectin 8
Test Time 1 hrs 20 mins
Total Assay Time 4 hrs 45 mins
Show More Show Less