missing translation for 'onlineSavingsMsg'
Learn More

Human Galectin-3 Alexa Fluor 594-conjugated Antibody, R&D Systems™

Catalog No. IC11542T100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
IC11542T100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. IC11542T100 Supplier Bio-Techne Supplier No. IC11542T100UG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Mouse Monoclonal Antibody

Galectin-3 Monoclonal specifically detects Galectin-3 in Human samples. It is validated for Flow Cytometry (Intracellular Staining).

Specifications

Antigen Galectin-3
Applications Flow Cytometry (Intracellular Staining)
Classification Monoclonal
Clone 194801
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 594
Dilution Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Accession No. P17931
Gene Alias AGE-R3, Carbohydrate-binding protein 35, CBP 35,35 kDa lectin, CBP35, Gal-3, GAL3, galactin-3, Galactose-specific lectin 3, Galactoside-binding protein, GALBPCBP35, galectin-3, Galectin3, GALIG, IgE-binding protein, L29, L31, L-31, Laminin-binding protein, Lectin L-29, lectin, galactoside-binding, soluble, 3, LGALS2, LGALS3, Mac-2, Mac-2 antigen, MAC2GAL3
Gene Symbols LGALS3
Host Species Mouse
Immunogen E. coli-derived recombinant human Galectin-3 Ala2-Ile250 Accession # P17931
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3958
Test Specificity Detects human Galectin-3 in direct ELISAs. In direct ELISAs, 100% cross-reactivity with recombinant mouse Galectin-3 is observed and no cross-reactivity with recombinant human (rh) Galectin-8 or rhGalectin-9 is observed.
Target Species Human
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Product Type Antibody
Isotype IgG2b
Product Line R&D Systems
Show More Show Less

For Research Use Only (RUO)