missing translation for 'onlineSavingsMsg'
Learn More

Human Galanin Alexa Fluor 405-conjugated Antibody, R&D Systems™

Catalog No. IC5854V100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
IC5854V100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. IC5854V100 Supplier Bio-Techne Supplier No. IC5854V100UG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Mouse Monoclonal Antibody

Galanin Monoclonal specifically detects Galanin in Human samples. It is validated for Flow Cytometry (Intracellular Staining).

Specifications

Antigen Galanin
Applications Flow Cytometry (Intracellular Staining)
Classification Monoclonal
Clone 581403
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 405
Dilution Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Accession No. P22466
Gene Alias GAL, GAL1, galanin, galanin prepropeptide, GALN, GALNgalanin-related peptide, GLNNgalanin-message-associated peptide, GMAP, MGC40167
Gene Symbols GAL
Host Species Mouse
Immunogen E.coli-derived recombinant human Galanin Ala20-Ser123 Accession # P22466
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51083
Test Specificity Detects human Galanin in direct ELISAs.
Target Species Human
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Product Type Antibody
Isotype IgG2b
Product Line R&D Systems
Show More Show Less

For Research Use Only (RUO)