missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human Fructosamine-3-kinase (NP_071441.1) VersaClone cDNA

Specifications
Specifications
| Gene | FN3K |
| Accession Number | NP_071441.1 |
| Gene ID (Entrez) | 64122 |
| Gene Alias | EC 2.7.1.-, FN3K, fructosamine 3 kinase, fructosamine-3-kinase |
| Content And Storage | Store the unopened product at -20 to -70 °C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Species | Human |
| Product Line | R&D Systems |
| Quantity | 10 μg |