missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human FOXR1 (aa 146-284) Control Fragment Recombinant Protein

Catalog No. RP109716
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP109716 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP109716 Supplier Invitrogen™ Supplier No. RP109716
Only null left

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (54%), Rat (54%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-144774 (PA5-144774. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

The forkhead-box (FOX) genes comprise a superfamily of at least 43 members that express proteins which are involved in transcriptional regulation and may be associated with the pathogenesis of various cancers. FOX family members are monomeric, helix-turn-helix proteins with a core DNA-binding domain of approximately 110 amino acids. FOX transcription factors play roles in determining cell fate during early development. FOXR1 (forkhead box protein R1), also known as FOXN5 (forkhead box protein N5) or DLNB13, is a 292 amino acid nuclear protein that belongs to the FOX family and contains one forkhead DNA-binding domain. Through chromosomal aberrations such as retroviral integration, gene amplification or translocation, FOXR1 may be involved the development of certain invasive carcinomas.

Specifications

Accession Number Q6PIV2
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 283150
Name Human FOXR1 (aa 146-284) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias DLNB13; Forkhead box protein N5; forkhead box protein R1; forkhead box R1; forkhead box R1 variant 1; forkhead box R1 variant 2; Foxn5; FOXR1; Gm1115
Common Name FOXR1
Gene Symbol Foxr1
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence LPSPHKRAPLQSRRLRQASSQAGRLWSRPPLNYFHLIALALRNSSPCGLNVQQIYSFTRKHFPFFRTAPEGWKNTVRHNLCFRDSFEKVPVSMQGGASTRPRSCLWKLTEEGHRRFAEEARALASTRLESIQQCMSQPD
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less