missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human FOLR2 ELISA Kit
ELISA
Supplier: Invitrogen™ EH200RB
Description
Human FOLR2 quantitates human FOLR2 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human FOLR2.
FOLR2 is a member of the folate receptor (FOLR) family, and these genes exist in a cluster on chromosome 11. Members of this gene family have a high affinity for folic acid and for several reduced folic acid derivatives, and they mediate delivery of 5-methyltetrahydrofolate to the interior of cells. This protein has a 68% and 79% sequence homology with the FOLR1 and FOLR3 proteins, respectively. Although this protein was originally thought to be specific to placenta, it can also exist in other tissues, and it may play a role in the transport of methotrexate in synovial macrophages in rheumatoid arthritis patients.Chemical Identifiers
| BETA-HFR; FBP; FBP/PL-1; FBP2; folate binding protein 2; Folate receptor 2; folate receptor 2 (fetal); folate receptor beta; folate receptor, beta; folate receptor, fetal/placental; Folate-binding protein 2; folate-binding protein, fetal/placental; Folbp2; Folbp-2; FOLR2; FR-beta; FR-P3; placental folate-binding protein | |
| 0.17 ng/mL | |
| ELISA | |
| Colorimetric Microplate Reader | |
| FOLR2 | |
| <10 | |
| HRP | |
| Plasma 50 μL, Serum 50 μL, Supernatant 50 μL | |
| FOLR2 | |
| 4 hrs 45 mins |
| 0.17 to 10 ng/mL | |
| HRP | |
| Plasma, Serum, Supernatant | |
| 2350 | |
| <12% | |
| Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
| RUO | |
| 2°C to 8°C | |
| 1 hrs 20 mins |
Specifications
| BETA-HFR; FBP; FBP/PL-1; FBP2; folate binding protein 2; Folate receptor 2; folate receptor 2 (fetal); folate receptor beta; folate receptor, beta; folate receptor, fetal/placental; Folate-binding protein 2; folate-binding protein, fetal/placental; Folbp2; Folbp-2; FOLR2; FR-beta; FR-P3; placental folate-binding protein | |
| 0.17 ng/mL | |
| ELISA | |
| Colorimetric Microplate Reader | |
| FOLR2 | |
| <10 | |
| HRP | |
| RUO | |
| 2°C to 8°C | |
| 1 hrs 20 mins |
| 0.17 to 10 ng/mL | |
| HRP | |
| Plasma, Serum, Supernatant | |
| 2350 | |
| <12% | |
| Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
| 96 Tests | |
| Plasma 50 μL, Serum 50 μL, Supernatant 50 μL | |
| FOLR2 | |
| 4 hrs 45 mins |