missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human Fibrinogen Native Protein

Catalog No. PIRP43142
Change view
Click to view available options
Quantity:
2 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIRP43142 2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIRP43142 Supplier Invitrogen™ Supplier No. RP43142

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Native Protein

RP-43142 contains native human Fibrinogen protein. RP-43142 may be used as a positive control in many immunological assays. The product is of human blood/plasma origin. Although the starting material was tested prior to initiation of the manufacturing process, and was found negative or nonreactive for anti-HIV-1/2, HIV-1 antigen(s), HBsAg, STS, anti-HCV, anti-HBcore and anti-HTLV I & II, extreme caution should be used when handling this material as there is a margin of error in all tests. This product is to be used for IN VITRO research purposes only, and is not intended for clinical or diagnostic use!.

Fibrinogen, the precursor of fibrin, is the coagulable protein in the blood plasma of vertebrates. Following vascular injury, fibrinogen is cleaved by thrombin to form fibrin which is the most abundant component of blood clots. Fibrinogen consists as a dimer of 3 pairs of non-identical chains FGA, FGB, and FGG that are cross-linked by disulfide bonds in their N-terminal segments. The molecule has 2 terminal D domains and one central E domain, all three domains are separated when fibrinogen is degradated by plasmin. Mutations in this gene lead to several disorders, including dysfibrinogenemia, afibrinogenemia, and renal amyloidosis.

Specifications

Accession Number P02671, P02675, P02679
Concentration 4.3 mg/mL
For Use With (Application) Neutralization
Formulation 0.01M sodium citrate, 0.01M sodium phosphate with no preservative; pH 7.3
Gene ID (Entrez) 2243, 2244, 2266
Molecular Weight (g/mol) 340 kDa
Name Human Fibrinogen
pH Range 7.3
Purification Method SDS-PAGE
Quantity 2 mg
Source Human
Storage Requirements -80°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias 2510049G14Rik; 3010002H13Rik; A-alpha fibrinogen; Ab1-181; Ab1-216; Ac1-581; Ac1873; AI256424; carboxy-terminal region; ENSMUSG00000059807; epididymis secretory sperm binding protein Li 78p; Fba5e; Fga; Fgb; Fgg; Fib; Fib2; fibg; fibrinogen; fibrinogen A-alpha chain; fibrinogen A-alpha-chain; fibrinogen alpha chain; fibrinogen alpha-E subunit; fibrinogen beta chain; Fibrinogen gamma chain; fibrinogen gamma polypeptide; fibrinogen gamma-B chain; fibrinogen gamma-chain; fibrinogen, A alpha polypeptide; fibrinogen, alpha polypeptide; fibrinogen, B beta polypeptide; fibrinogen, beta polypeptide; fibrinogen, gamma polypeptide; Fibrinopeptide A; Fibrinopeptide B; Gamma'; gamma-fibrinogen; HEL-S-78p; Liver regeneration-related protein LRRG036/LRRG043/LRRG189; MGC119422; MGC119423; MGC119425; PRO2061; testicular tissue protein Li 70; Unknown (protein for MGC:127951)
Common Name Fibrinogen
Gene Symbol FGA, FGB, FGG
Product Type Protein
Conjugate Unconjugated
Species Human
Content And Storage -80°C, Avoid Freeze/Thaw Cycles
Form Liquid
Protein Subtype Native
Show More Show Less