missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human Fibrinogen Fragment E Native Protein

Catalog No. PIRP43144
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIRP43144 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIRP43144 Supplier Invitrogen™ Supplier No. RP43144

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Native Protein

RP-43144 contains native human Fibrinogen Fragment E protein and less than 1% of Fragment D. RP-43144 may be used as a positive control in many immunological assays. The product is of human blood/plasma origin. Although the starting material was tested prior to initiation of the manufacturing process, and was found negative or nonreactive for anti-HIV-1/2, HIV-1 antigen(s), HBsAg, STS, anti-HCV, anti-HBcore and anti-HTLV I & II, extreme caution should be used when handling this material as there is a margin of error in all tests. This product is to be used for IN VITRO research purposes only, and is not intended for clinical or diagnostic use! Reconstitute with 606 mL of water to achieve 3.3 mg/mL.

Specifications

Accession Number P02671
Concentration 0.5 mg/mL
For Use With (Application) Neutralization
Formulation Protein with 0.9% NaCl, 3% glycine and no preservative; pH 7.4
Gene ID (Entrez) 2243
Molecular Weight (g/mol) 50 kDa
Name Human Fibrinogen Fragment E
pH Range 7.4
Purification Method SDS-PAGE
Quantity 100 μg
Source Human
Storage Requirements 4°C
Regulatory Status RUO
Gene Alias FGA; Fib2; fibrinogen alpha chain; fibrinogen, A alpha polypeptide; Fibrinopeptide A
Common Name Fibrinogen Fragment E
Gene Symbol FGA
Product Type Protein
Conjugate Unconjugated
Species Human
Content And Storage 4°C
Form Lyophilized
Protein Subtype Native
Show More Show Less