Learn More
Invitrogen™ Human FGF2 ELISA Development Kit (ABTS), PeproTech®
ELISA
Supplier:  Invitrogen™ 900K08K
Catalog No. 90-0K0-8K
Only null left
Explore available promotions
Description
Description
FGF2 (FGFb, fibroblast growth factor basic) belongs to the fibroblast growth factor (FGF) family, and interacts with high-affinity transmembrane receptors to influence cell proliferation and tissue neovascularization. FGF2 exists as five isoforms with distinct intracellular localizations and functions. The 18 kDa isoform is predominantly cytosolic and acts through cell surface receptors, whereas the 22, 22.5, 24 and 34 kDa isoforms are nuclear and may signal independent of transmembrane receptor pathways. In humans, the gene is located on the q arm of chromosome 4. FGF2 has been implicated in diverse biological processes, such as limb and nervous system development, wound healing, and tumor growth. The mRNA for FGF2 contains multiple polyadenylation sites, and is alternatively translated from non-AUG and AUG initiation codons, resulting in five different isoforms with distinct properties. The CUG-initiated isoforms are localized in the nucleus and are responsible for the intracrine effect, whereas, the AUG-initiated form is mostly cytosolic and is responsible for the paracrine and autocrine effects of this FGF. Diseases associated with FGF2 dysfunction include Kaposi Sarcoma and corneal neovascularization.
Specifications
Specifications
P09038
Lot-specific
ELISA
2247
Capture Antibody, Detection Antibody, Protein Standard, HRP-conjugate
10 x 96 Tests
Refer to product documentation for component specific storage temperature.
1 hrs 45 mins
Lot-specific
HRP
Colorimetric Microplate Reader
FGF2
HRP
RUO
FGF2
24 hrs 30 mins
Product Suggestions
Product Suggestions