Learn More
Gibco™ Human FGF-basic (FGF-2/bFGF) (aa 10-155) Recombinant Protein DFS Item
Recombinant Protein
Supplier:  Gibco™ PHG0021L
Catalog No. PHG0021L
Only null left
Explore available promotions
Description
Description
Carrier-Free Reconstitution: Upon initial thawing, centrifuge the vial briefly, before opening to bring the contents to the bottom. Apportion the protein into working aliquots and store at ≤-20°C. Make any further dilutions of the reconstituted protein in low endotoxin medium or buffered solution with FBS or tissue culture grade BSA. Storage: Store the unopened vial at ≤-20°C. Upon thawing, apportion into working aliquots and store at ≤-20°C. Avoid repeated freeze-thaw cycles.
FGF2 (FGFb, fibroblast growth factor basic) belongs to the fibroblast growth factor (FGF) family, and interacts with high-affinity transmembrane receptors to influence cell proliferation and tissue neovascularization. FGF2 exists as five isoforms with distinct intracellular localizations and functions. The 18 kDa isoform is predominantly cytosolic and acts through cell surface receptors, whereas the 22, 22.5, 24 and 34 kDa isoforms are nuclear and may signal independent of transmembrane receptor pathways. In humans, the gene is located on the q arm of chromosome 4. FGF2 has been implicated in diverse biological processes, such as limb and nervous system development, wound healing, and tumor growth. The mRNA for FGF2 contains multiple polyadenylation sites, and is alternatively translated from non-AUG and AUG initiation codons, resulting in five different isoforms with distinct properties. The CUG-initiated isoforms are localized in the nucleus and are responsible for the intracrine effect, whereas, the AUG-initiated form is mostly cytosolic and is responsible for the paracrine and autocrine effects of this FGF. Diseases associated with FGF2 dysfunction include Kaposi Sarcoma and corneal neovascularization.
Specifications
Specifications
P09038
20mM citrate with no preservative; pH 5
17 kDa
5
100 μg
RUO
Basic fibroblast growth factor; basic fibroblast growth factor bFGF; bFGF; FGF; fgf basic; Fgf2; Fgf-2; Fgfb; FGF-b; Fibroblast growth factor; fibroblast growth factor 2; fibroblast growth factor 2 (basic); Fibroblast growth factor basic; HBGF-2; Heparin-binding growth factor 2; H-FGF-b-147; H-FGF-b-154; M-FGF-b; Prostatic growth factor; prostatropin
FGF2
Protein
Human
PALPEDGGSG AFPPGHFKDP KRLYCKNGGF FLRIHPDGRV DGVREKSDPH IKLQLQAEER GVVSIKGVCA NRYLAMKEDG RLLASKCVTD ECFFFERLES NNYNTYRSRK YTSWYVALKR TGQYKLGSKT GPGQKAILFL PMSAKS
ED50 ≤ 1.0 ng/mL; determined by the dose-dependent proliferation of BALB/3T3 cells.
Balb/3T3 cells
1.0 x 107 - 1.0 x 106 units⁄mg
Growth Factors & Receptors
Lyophilized
Neurobiology, Oncology, Stem Cell Research, Microbiology, Angiogenesis
≥95% by SDS-PAGE
Bioactivity
2247
Human FGF-basic (FGF-2/bFGF) (aa 10-155)
Purified
-20°C
<0.1 ng/μg
FGF2
ED50 ≤ 1.0 ng/mL; determined by the dose-dependent proliferation of BALB/3T3 cells.
Unconjugated
Recombinant
-20°C
<0.1 ng/μg
Dry Ice
E. coli
Recombinant, Ligand, Truncated
FGFs (Fibroblast Growth Factors)
Gibco™
Product Suggestions
Product Suggestions