missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Human FGF-6 Recombinant Protein, PeproTech®
Recombinant Protein
$132.00 - $4890.00
Specifications
| Accession Number | P10767 |
|---|---|
| For Use With (Application) | Functional Assay |
| Formulation | protein with no preservative |
| Gene ID (Entrez) | 2251 |
| Molecular Weight (g/mol) | 18.7 kDa |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
10030100UG
|
Gibco™
10030100UG |
100 μg |
Each for $931.06
|
|
|||||
100301MG
|
Gibco™
LS100301MG |
1 mg |
Each for $4,890.00
|
|
|||||
10030250UG
|
Gibco™
10030250UG |
250 μg |
Each for $1,628.00
|
|
|||||
1003025UG
|
Gibco™
LS1003025UG |
25 μg |
Each for $318.00
|
|
|||||
10030500UG
|
Gibco™
10030500UG |
500 μg |
Each for $2,795.00
|
|
|||||
100305UG
|
Gibco™
LS100305UG |
5 μg |
Each for $132.00
|
|
|||||
Description
Recombinant Human FGF-6 is an 18.7 kDa protein consisting of 168 amino acid residues. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis
FGF-6 is a secreted, heparin-binding growth factor that is a member of the FGF family. Proteins of this family play a central role during prenatal development, postnatal growth and regeneration of a variety of tissues, by promoting cellular proliferation and differentiation. FGF-6 is expressed in leukemia cell lines with platelet megakaryocytic differentiation potential. It signals through FGFR 1c, 2c, and 4.4Specifications
| P10767 | |
| protein with no preservative | |
| 18.7 kDa | |
| E. coli | |
| <1 EU/ μg | |
| FGF6 | |
| Determined by a cell proliferation assay using Balb/c 3T3 cells. The expected ED50 is ≤ 0.1 ng/ml, corresponding to a specific activity of ≥ 1 x 10^7 units/mg | |
| Recombinant | |
| -20°C | |
| Lyophilized |
| Functional Assay | |
| 2251 | |
| Human FGF-6 | |
| RUO | |
| FGF; FGF6; Fgf-6; Fibroblast growth factor; Fibroblast growth factor 6; HBGF-6; Heparin secretory-transforming protein 2; heparin-binding growth factor 6; HST2; HST-2; Hstf2; HSTF-2 | |
| FGF6 | |
| Unconjugated | |
| GTRANNTLLD SRGWGTLLSR SRAGLAGEIA GVNWESGYLV GIKRQRRLYC NVGIGFHLQV LPDGRISGTH EENPYSLLEI STVERGVVSL FGVRSALFVA MNSKGRLYAT PSFQEECKFR ETLLPNNYNA YESDLYQGTY IALSKYGRVK RGSKVSPIMT VTHFLPRI | |
| E. coli | |
| ≥ 95% by SDS-PAGE gel and HPLC analyses. |