missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Human FGF-4, Animal-Free Recombinant Protein, PeproTech®
Recombinant Protein
Supplier: Gibco™ AF1003125UG
Description
AF-100-31-1MG will be provided as 2 x 500 μg (AF-100-31-500UG). Recombinant Human FGF-4 is a 19.7 kDa protein consisting of 182 amino acid residues. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis
Fibroblast growth factor 4 (FGF4) is a member of the fibroblast growth factor (FGF) family that possess broad mitogenic and cell survival activities and play key roles in growth and survival of stem cells during embryogenesis, tissue regeneration, and carcinogenesis. FGF4 was identified by its strong oncogenic transforming activity and is a potent angiogenic factor, expressed in several highly vascularized tumors and also in adult mouse testis, intestine, and brain. Studies on the mouse homolog suggests a function in bone morphogenesis and limb development through the sonic hedgehog (SHH) signaling pathway. Furthermore, FGF4 regulates neural progenitor cell proliferation and neuronal differentiation. Recent studies show a growth-promoting role for FGF4 in human embryonic stem cells and a putative feedback inhibition mechanism by a novel FGF4 splice isoform that may serve to promote differentiation at a later stages of development.Specifications
| P08620 | |
| protein with no preservative | |
| 19.7 kDa | |
| 25 μg | |
| RUO | |
| FGF; FGF4; Fgf-4; Fgfk; Fibroblast growth factor; fibroblast growth factor 4; fibroblast growth factor 4 splice isoform; HBGF-4; heparin secretory transforming protein 1; heparin secretory-transforming protein 1; heparin-binding growth factor 1; heparin-binding growth factor 4; HST; Hst1; hst-1; HSTF1; HSTF-1; human stomach cancer, transforming factor from FGF-related oncogene; kaposi sarcoma oncogene; Kfgf; K-FGF; K-fibroblast growth factor; KS3; oncogene HST; transforming protein KS3 | |
| Fgf4 | |
| Unconjugated | |
| GRGGAAAPTA PNGTLEAELE RRWESLVALS LARLPVAAQP KEAAVQSGAG DYLLGIKRLR RLYCNVGIGF HLQALPDGRI GGAHADTRDS LLELSPVERG VVSIFGVASR FFVAMSSKGK LYGSPFFTDE CTFKEILLPN NYNAYESYKY PGMFIALSKN GKTKKGNRVS PTMKVTHFLP RL | |
| E. coli | |
| ≥ 95% by SDS-PAGE gel and HPLC analyses. |
| Functional Assay | |
| 2249 | |
| Human FGF-4, Animal-Free | |
| E. coli | |
| <0.1 EU/ μg | |
| FGF4 | |
| Determined by a cell proliferation assay using Balb/c 3T3 cells. The expected ED50 is ≤ 10 ng/ml, corresponding to a specific activity of ≥ 1 x 10^5 units/mg | |
| Recombinant | |
| -20°C | |
| Lyophilized |