missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human FGF-21 ELISA Kit
ELISA
$890.00 - $4,420.00
Specifications
| Accession Number | FGF; FGF21; FGF-21; Fibroblast growth factor; fibroblast growth factor 21; UNQ3115/PRO10196 |
|---|---|
| Assay Range | 8 to 2,000 pg/mL |
| Assay Sensitivity | 8 pg/mL |
| Conjugate | HRP |
| Product Type | ELISA |
Description
Human FGF-21 quantitates human FGF-21 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human FGF-21.
The protein encoded by this gene is a member of the fibroblast growth factor family. FGF family members possess broad mitogenic and cell survival activities and are involved in a variety of biological processes including embryonic development, cell growth, morphogenesis, tissue repair, tumor growth and invasion. The function of this growth factor has not yet been determined.Specifications
| FGF; FGF21; FGF-21; Fibroblast growth factor; fibroblast growth factor 21; UNQ3115/PRO10196 | |
| 8 pg/mL | |
| ELISA | |
| Colorimetric Microplate Reader | |
| FGF21 | |
| <10 | |
| HRP | |
| Plasma 50 μL, Serum 50 μL, Supernatant 50 μL | |
| FGF21 | |
| 4 hrs 45 mins |
| 8 to 2,000 pg/mL | |
| HRP | |
| Plasma, Serum, Supernatant | |
| 26291 | |
| <12% | |
| Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
| RUO | |
| 2°C to 8°C | |
| 1 hrs 20 mins |