missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Human FGF-16 Recombinant Protein, PeproTech®
Recombinant Protein
$132.00 - $11551.52
Specifications
| Accession Number | O43320 |
|---|---|
| For Use With (Application) | ELISA, Functional Assay, Western Blot |
| Formulation | protein with no preservative |
| Gene ID (Entrez) | 8823 |
| Name | Human FGF-16 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
10029100UG
|
Gibco™
10029100UG |
100 μg |
Each for $931.06
|
|
|||||
100291MG
|
Gibco™
LS100291MG |
4 x 250 μg |
Each for $4,630.00
|
|
|||||
|
10-029-01M
|
Gibco™
1002901M |
4 x 250 μg |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
10029250UG
|
Gibco™
10029250UG |
250 μg |
Each for $1,628.00
|
|
|||||
|
10-029-500
|
Gibco™
10029500 |
2 x 250 μg |
Each for $9,455.81
|
|
|||||
1002925UG
|
Gibco™
LS1002925UG |
25 μg |
Each for $318.00
|
|
|||||
10029500UG
|
Gibco™
10029500UG |
2 x 250 μg |
Each for $2,780.00
|
|
|||||
100295UG
|
Gibco™
LS100295UG |
5 μg |
Each for $132.00
|
|
|||||
Description
100-29-1MG will be provided as 4 x 250 μg (100-29-250UG). 100-29-500UG will be provided as 2 x 250 μg (100-29-250UG). Recombinant Human FGF-16 is a 23.6 kDa protein consisting of 206 amino acid residues. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis
FGF-16 is a heparin-binding growth factor that is a member of the FGF family. Proteins of this family play a central role during prenatal development, postnatal growth and regeneration of a variety of tissues, by promoting cellular proliferation and differentiation. FGF-16 signals through FGFR 2c and 3c. FGF-16 plays a role in the development of the central nervous system.Specifications
| O43320 | |
| protein with no preservative | |
| Human FGF-16 | |
| RUO | |
| FGF16 | |
| Recombinant | |
| Lyophilized |
| ELISA, Functional Assay, Western Blot | |
| 8823 | |
| E. coli | |
| FGF; FGF16; FGF-16; Fibroblast growth factor; Fibroblast growth factor 16; metacarpal 4-5 fusion; MF4 | |
| Unconjugated | |
| AEVGGVFASL DWDLHGFSSS LGNVPLADSP GFLNERLGQI EGKLQRGSPT DFAHLKGILR RRQLYCRTGF HLEIFPNGTV HGTRHDHSRF GILEFISLAV GLISIRGVDS GLYLGMNERG ELYGSKKLTR ECVFREQFEE NWYNTYASTL YKHSDSERQY YVALNKDGSP REGYRTKRHQ KFTHFLPRPV DPSKLPSMSR DLFHYR | |
| ≥ 95% by SDS-PAGE gel and HPLC analyses. |