missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Human FGF-10 Recombinant Protein, PeproTech®
Recombinant Protein
Supplier: Gibco™ LS100261MG
Description
100-00CC-500UG will be provided as 2 x 250 μg (100-00CC-250UG). Recombinant Human FGF-10 is a 19.3 kDa protein consisting of 170 amino acid residues. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis
FGF10 is a member of the fibroblast growth factor (FGF) family. FGF family members possess broad mitogenic and cell survival activities, and are involved in a variety of biological processes, including embryonic development, cell growth, morphogenesis, tissue repair, tumor growth and invasion. This protein exhibits mitogenic activity for keratinizing epidermal cells, but essentially no activity for fibroblasts, which is similar to the biological activity of FGF7. Studies of the mouse homolog of suggested that this gene is required for embryonic epidermal morphogenesis including brain development, lung morphogenesis, and initiation of lim bud formation. This protein is also implicated to be a primary factor in the process of wound healing.Specifications
| O15520 | |
| protein with no preservative | |
| 19.3 | |
| 2 x 500 μg | |
| -20°C | |
| <1 EU/μg | |
| FGF10 | |
| Unconjugated | |
| MLGQDMVSPE ATNSSSSSFS SPSSAGRHVR SYNHLQGDVR WRKLFSFTKY FLKIEKNGKV SGTKKENCPY SILEITSVEI GVVAVKAINS NYYLAMNKKG KLYGSKEFNN DCKLKERIEE NGYNTYASFN WQHNGRQMYV ALNGKGAPRR GQKTRRKNTS AHFLPMVVHS | |
| ≥ 95% by SDS-PAGE gel and HPLC analyses. |
| ELISA, Functional Assay, Western Blot | |
| 2255 | |
| Human FGF-10 | |
| E. coli | |
| RUO | |
| AEY17; BB213776; FGF; Fgf10; FGF-10; Fibroblast growth factor; Fibroblast growth factor 10; Gsfaey17; Keratinocyte growth factor 2; produced by fibroblasts of urinary bladder lamina propria | |
| Human | |
| Recombinant | |
| Lyophilized |