missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human Fas Ligand/TNFSF6 Quantikine ELISA Kit

Catalog No. DFL00B
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DFL00B 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. DFL00B Supplier R&D Systems™ Supplier No. DFL00B

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Assay Range 15.60 - 1000 pg/mL
Assay Sensitivity 5.1 pg/mL
Product Line R&D Systems
Sample Type Cell Culture Supernatant, Cell Lysates, Serum, Plasma (EDTA), Plasma (Heparin)
For Use With (Application) ELISA
Gene ID (Entrez) 356
Quantity 1 Kit
Storage Requirements Store the unopened product at 2 - 8°C. Do not use past expiration date.
Synonym apoptosis (APO-1) antigen ligand 1, Apoptosis antigen ligand, APT1LG1CD95L, APTL, CD178, CD178 antigen, CD95L, CD95-L, Fas antigen ligand, Fas ligand, Fas ligand (TNF superfamily, member 6), FASLCD95 ligand, FASLG, TNFSF6, TNFSF6FasL, tumor necrosis factor (ligand) superfamily, member 6, tumor necrosis factor ligand superfamily member 6
Target Human