missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human FAM83H (aa 1085-1175) Control Fragment Recombinant Protein

Catalog No. RP93935
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP93935 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP93935 Supplier Invitrogen™ Supplier No. RP93935
Only null left

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (89%), Rat (89%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-55094 (PA5-55094. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

The protein encoded by this gene plays an important role in the structural development and calcification of tooth enamel. Defects in this gene are a cause of amelogenesis imperfecta type 3 (AI3).

Specifications

Accession Number Q6ZRV2
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 286077
Name Human FAM83H (aa 1085-1175) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias AA409316; AI3; FAM83H; FAM83H variant 1; FAM83H variant 2; family with sequence similarity 83 member H; family with sequence similarity 83, member H; FLJ46072; protein FAM83H; RGD1305866
Common Name FAM83H
Gene Symbol FAM83H
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence SDKDKCSAIFRSDSLGTQGRLSRTLPASAEERDRLLRRMESMRKEKRVYSRFEVFCKKEEASSPGAGEGPAEEGTRDSKVGKFVPKILGTF
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less