missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human FAM81A (aa 77-216) Control Fragment Recombinant Protein

Catalog No. RP101031
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP101031 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP101031 Supplier Invitrogen™ Supplier No. RP101031

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (99%), Rat (99%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-51644 (PA5-51644. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

FAM81A (Family With Sequence Similarity 81 Member A) is a Protein Coding gene. An important paralog of this gene isFAM81B.

Specifications

Accession Number Q8TBF8
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 145773
Name Human FAM81A (aa 77-216) Control Fragment
Quantity 100 μL
Gene Alias 6430514L14Rik; FAM81A; family with sequence similarity 81 member A; family with sequence similarity 81, member A; protein FAM81A; RGD1311958
Common Name FAM81A
Gene Symbol FAM81A
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence LFLEEHIRNITAIVKQLNRDIEVLQEQIRARDNISYGTNSALKTLEMRQLSGLGDLRGRVARCDASIARLSAEHKTTYEGLQHLNKEQQAAKLILETKIKDAEGQISQLLNRVDLSISEQSTKLKMSHRDSNHQLQLLDT
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less