missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human FAM54A (aa 3-91) Control Fragment Recombinant Protein

Catalog No. RP94896
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP94896 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP94896 Supplier Invitrogen™ Supplier No. RP94896

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (71%), Rat (71%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-56259. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

FAM54A belongs to the MTFR1/FAM54 family. The function of the FAM54A protein is not known.

Specifications

Accession Number Q6P444
Concentration ≥5.0 mg/mL
For Use With (Application) Neutralization, Control
Formulation PBS, 1M urea with no preservative; pH 7.4
Gene ID (Entrez) 113115
Name Human FAM54A (aa 3-91) Control Fragment
pH Range 7.4
Purification Method Purified
Quantity 100 μL
Storage Requirements -20°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias DUF729 domain containing 1; DUF729 domain-containing protein 1; DUFD1; EGK_15321; FAM54A; family with sequence similarity 54, member A; LOC100911069; Mitochondrial fission regulator 2; Mtfr2; protein FAM54A; protein FAM54A-like
Common Name FAM54A
Gene Symbol MTFR2
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence LILNILREMLEYFGVPVEQVLLIWENKDYGSTRSIVRIIGKMLPLEPCRRPNFELIPLLNSVDSDNCGSMVPSFADILYVANDEEASYL
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less