missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human FAM177A1 (aa 153-212) Control Fragment Recombinant Protein
Recombinant Protein
Supplier: Invitrogen™ RP101372
Description
Highest antigen sequence indentity to the following orthologs: Mouse (83%), Rat (83%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-62762 (PA5-62762. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
This gene encodes a member of a conserved protein family. Alternative splicing results in multiple transcript variants. This gene is thought to be associated with susceptibility to juvenile idiopathic arthritis.Specifications
| Q8N128 | |
| Blocking Assay, Control | |
| 283635 | |
| 100 μL | |
| C14orf24; FAM177A1; family with sequence similarity 177 member A1; family with sequence similarity 177, member A1; protein FAM177A1 | |
| FAM177A1 | |
| Human | |
| His-ABP-tag | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ≥5.0 mg/mL | |
| 1 M urea, PBS with no preservative; pH 7.4 | |
| Human FAM177A1 (aa 153-212) Control Fragment | |
| RUO | |
| FAM177A1 | |
| Unconjugated | |
| Recombinant | |
| NRMSEEAEKQYQQNKLQTDSIVQTDQPETVISSSFVNVNFEMEGDSEVIMESKQNPVSVP | |
| E. coli | |
| >80% by SDS-PAGE and Coomassie blue staining |