missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human FAM173A (aa 123-218) Control Fragment Recombinant Protein

Catalog No. RP98370
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP98370 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP98370 Supplier Invitrogen™ Supplier No. RP98370
Only null left

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (74%), Rat (74%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-61198 (PA5-61198. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Specifications

Accession Number Q9BQD7
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 65990
Name Human FAM173A (aa 123-218) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias Adenine nucleotide translocase lysine N-methyltransferase; ANTKMT; ANT-KMT; C16orf24; FAM173A; family with sequence similarity 173 member A; family with sequence similarity 173, member A; protein FAM173A; RJD7
Common Name FAM173A
Gene Symbol FAM173A
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence CAGSVCYRRKDLWKVSLRDCRNVSVFLAPSVLPLLEDKLRTELPAGARVVSGRFPLPTWQPVTAVGEGLDRVWAYDVPEGGQAGEAASSRIPIQAA
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less