missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human FAM162B (aa 135-162) Control Fragment Recombinant Protein

Catalog No. RP95019
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP95019 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP95019 Supplier Invitrogen™ Supplier No. RP95019
Only null left

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (93%), Rat (93%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-57523 (PA5-57523. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Specifications

Accession Number Q5T6X4
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 221303
Name Human FAM162B (aa 135-162) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias bA86F4.2; C6orf189; f162; FAM162B; family with sequence similarity 162 member B; family with sequence similarity 162, member B; protein FAM162B
Common Name FAM162B
Gene Symbol FAM162B
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence HESLTSWNLAKKAKWREEAALAAQAKAK
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less