missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human FAM133B (aa 15-44) Control Fragment Recombinant Protein

Catalog No. RP105656
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP105656 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP105656 Supplier Invitrogen™ Supplier No. RP105656

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (100%), Rat (100%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-67053. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

FAM133B (Family With Sequence Similarity 133 Member B) is a Protein Coding gene. An important paralog of this gene isFAM133A.

Specifications

Accession Number Q5BKY9
Concentration ≥5.0 mg/mL
For Use With (Application) Neutralization, Control
Formulation PBS, 1M urea with no preservative; pH 7.4
Gene ID (Entrez) 257415
Name Human FAM133B (aa 15-44) Control Fragment
pH Range 7.4
Purification Method Purified
Quantity 100 μL
Storage Requirements -20°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias 2900022K02Rik; 5830415L20Rik; FAM133; Fam133b; family with sequence similarity 133 member B; family with sequence similarity 133, member B; LOC553430 protein; protein FAM133; Protein FAM133B; RGD1306302; si:ch211-287b5.3; Unknown (protein for IMAGE:8130787); wu:fc17e01; zgc:153800
Common Name FAM133B
Gene Symbol FAM133B
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence AMARSRGPIQSSGPTIQDYLNRPRPTWEEV
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less