missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human FAM110C (aa 102-152) Control Fragment Recombinant Protein

Catalog No. RP96215
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP96215 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP96215 Supplier Invitrogen™ Supplier No. RP96215

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (55%), Rat (55%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-57542 (PA5-57542. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

FAM110C may play a role in microtubule organization.

Specifications

Accession Number Q1W6H9
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 642273
Name Human FAM110C (aa 102-152) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias FAM110C; family with sequence similarity 110 member C; family with sequence similarity 110, member C; protein FAM110C
Common Name FAM110C
Gene Symbol FAM110C
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence IYRQKCEFVRGSGADGPRASLVKKLFQGPGKDKAPVPRTGDEGKAGNPETV
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less