missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human FAK (NP_722560.1) VersaClone cDNA

Specifications
Specifications
| Gene | FAK |
| Accession Number | NP_722560.1 |
| Gene ID (Entrez) | 5747 |
| Gene Alias | EC 2.7.10, FADK 1, FADK1, FADKFAK-related non-kinase polypeptide, FAK1FRNK, FAKEC 2.7.10.2, focal adhesion kinase 1, pp125FAK, Protein-tyrosine kinase 2, PTK2, PTK2 protein tyrosine kinase 2 |
| Content And Storage | Store the unopened product at -20 to -70 °C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Species | Human |
| Product Line | R&D Systems |
| Quantity | 10 μg |