missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human EXOC3L1 (aa 351-488) Control Fragment Recombinant Protein

Catalog No. RP92438
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP92438 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP92438 Supplier Invitrogen™ Supplier No. RP92438
Only null left

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (77%), Rat (77%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-56196 (PA5-56196. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

As part of the exocyst, may play a role in regulated exocytosis of insulin granules.

Specifications

Accession Number Q86VI1
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 283849
Name Human EXOC3L1 (aa 351-488) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias C730015A04Rik; EXOC3L; EXOC3L1; exocyst complex component 3 like 1; exocyst complex component 3-like; exocyst complex component 3-like 1; exocyst complex component 3-like protein; Protein Jiangli
Common Name EXOC3L1
Gene Symbol EXOC3L1
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence MGSLELGPEADVSQLEPLLTLENIEQLEATFVANIQASVSQWLQNALDGEVAEWGREHGPNTDPSGSYYSPMPAIVLQILEENIRVASLVSESLQQRVHGMALSELGTFLRSFSDALIRFSRDHFRGKSMAPHYVPYL
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less