missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human EVI5 (aa 719-810) Control Fragment Recombinant Protein

Catalog No. RP106490
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP106490 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP106490 Supplier Invitrogen™ Supplier No. RP106490

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (71%), Rat (71%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-65818 (PA5-65818. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

EVI5 is a novel centrosomal protein with a complex expression pattern and subcellular localization, possibly involved in centrosome stability and dynamics. It may act as an oncogene.

Specifications

Accession Number O60447
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 7813
Name Human EVI5 (aa 719-810) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias dJ846D11.1 (ecotropic viral integration site 5); ecotropic viral integration site 5; ecotropic viral integration site 5 protein; ecotropic viral integration site 5 protein homolog; EVI5; EVI-5; NB4S; Neuroblastoma stage 4 S gene protein
Common Name EVI5
Gene Symbol EVI5
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence QPPFDGIHIVNHLIGDDESFHSSDEDFIDNSLQETGVGFPLHGKSGSMSLDPAVADGSESETEDSVLETRESNQVVQKERPPRRRESYSTTV
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less