missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human EVA1B (aa 4-30) Control Fragment Recombinant Protein

Catalog No. RP98465
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP98465 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP98465 Supplier Invitrogen™ Supplier No. RP98465

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (100%), Rat (100%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-60410 (PA5-60410. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

EVA1B is a protein coding gene.

Specifications

Accession Number Q9NVM1
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 55194
Name Human EVA1B (aa 4-30) Control Fragment
Quantity 100 μL
Gene Alias 2610027C15Rik; C1orf78; eva-1 homolog B; eva-1 homolog B (C. elegans); EVA1B; Fam176b; family with sequence similarity 176, member B; protein eva-1 homolog B; Protein FAM176B; RGD1308876
Common Name EVA1B
Gene Symbol EVA1B
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence PRRDMELLSNSLAAYAHIRANPESFGL
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less