missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human ESPN (aa 785-848) Control Fragment Recombinant Protein

Recombinant Protein

Supplier:  Invitrogen™ RP94521

Catalog No. RP94521


Only null left
Explore available promotions

Description

Description

Highest antigen sequence indentity to the following orthologs: Mouse (89%), Rat (89%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-55941 (PA5-55941. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Multifunctional actin-bundling protein. Plays a major role in regulating the organization, dimensions, dynamics and signaling capacities of the actin filament-rich, microvillus-type specializations that mediate sensory transduction in variouS mechanosensory and chemosensory cells.
Specifications

Specifications

B1AK53
Blocking Assay, Control
83715
100 μL
actin cytoskeletal regulatory protein; autosomal recessive deafness type 36 protein; DFNB36; Ectoplasmic specialization protein; espin; Espn; je; jerker; Jerker, deafness locus; LP2654
Espn
Human
His-ABP-tag
-20°C, Avoid Freeze/Thaw Cycles
Liquid
≥5.0 mg/mL
1 M urea, PBS with no preservative; pH 7.4
Human ESPN (aa 785-848) Control Fragment
RUO
ESPN
Unconjugated
Recombinant
SMPAWRRDLLRKKLEEEREQKRKEEERQKQEELRREKEQSEKLRTLGYDESKLAPWQRQVILKK
E. coli
>80% by SDS-PAGE and Coomassie blue staining
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions