missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human Erythropoietin/EPO Quantikine ELISA Kit, RUO

Catalog No. SEPRU0B
Change view
Click to view available options
Quantity:
1 Kit
1 Pack (6 Plates)
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SEPRU0B 1 Pack (6 Plates)
DEPRU0B 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. SEPRU0B Supplier R&D Systems™ Supplier No. SEPRU0B
Only null left

Human Erythropoietin/EPO Quantikine ELISA Kit, RUO

Specifications

Specificity Natural and recombinant human Epo.
Target Species Human
Gene ID (Entrez) 2056
Quantity 1 Pack (6 Plates)
Storage Requirements Store the unopened product at 2 - 8 °C. Do not use past expiration date.
Synonym ECYT5, EP, EPO, epoetin, erythropoietin, MGC138142, MVCD2