missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human ERMN (aa 171-265) Control Fragment Recombinant Protein

Catalog No. RP96087
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP96087 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP96087 Supplier Invitrogen™ Supplier No. RP96087

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (66%), Rat (66%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-58327 (PA5-58327. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

ERMIN plays a role in cytoskeletal rearrangements during the late wrapping and/or compaction phases of myelinogenesis as well as in maintenance and stability of myelin sheath in the adult. ERMIN may play an important role in late-stage oligodendroglia maturation, myelin/Ranvier node formation during CNS development, and in the maintenance and plasticity of related structures in the mature CNS.

Specifications

Accession Number Q8TAM6
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 57471
Name Human ERMN (aa 171-265) Control Fragment
Quantity 100 μL
Gene Alias A330104H05Rik; AI854460; ermin; ermin, ERM-like protein; Ermn; JN; juxtanodin; Kiaa1189; mKIAA1189; RGD1308367
Common Name ERMN
Gene Symbol Ermn
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence LHSKHDEEQKVWDEEIDDDDDDNCNNDEDEVRVIEFKKKHEEVSQFKEEGDASEDSPLSSASSQAVTPDEQPTLGKKSDISRNAYSRYNTISYRK
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less