missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human Erc2 (aa 234-265) Control Fragment Recombinant Protein

Catalog No. RP108264
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP108264 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP108264 Supplier Invitrogen™ Supplier No. RP108264

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (100%), Rat (100%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-84939 (PA5-84939. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

ERC2 is a protein coding gene.

Specifications

Accession Number O15083
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 26059
Name Human Erc2 (aa 234-265) Control Fragment
Quantity 100 μL
Gene Alias 6430531D06; CAST; CAST1; CAST1/ERC2; CAZ-associated structural protein; CAZ-associated structural protein 1; Cmbp; cytomatrix protein p110; D14Ertd171e; ELKS/RAB6-interacting/CAST family member 2; ELKS2alpha; ELKSL; ERC protein 2; ERC2; KIAA0378; SPBC110; Spc110
Common Name Erc2
Gene Symbol Erc2
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence TQRDLNHLLQQESGNRGAEHFTIELTEENFRR
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less